Rabbit Polyclonal anti-Merlin [p Ser518] Antibody, Novus Biologicals (NBP1-51476)

NBP1-51476
102181-712EA 466.15 USD
102181-712
Rabbit Polyclonal anti-Merlin [p Ser518] Antibody, Novus Biologicals (NBP1-51476)
Rabbit Polyclonal Merlin [p Ser518] Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: Merlin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR