Anti-MEN1 Rabbit Polyclonal Antibody (Agarose immobilized)

NB600-421
102133-942EA 635.38 USD
102133-942
Anti-MEN1 Rabbit Polyclonal Antibody (Agarose immobilized)
Rabbit Polyclonal Menin Antibody [Agarose Immobilized]. Tested Applications: Immunoprecipitation. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MEN1
Clonality: Polyclonal
Clone:
Conjugation: Agarose immobilized
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR