|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | MTNR1B | |
Antigen Name | Melatonin Receptor 1B | |
Reactivity | Human, Rat, Mouse | Mouse, Rat, Human |
Conjugation | Unconjugated | |
Clonality | Polyclonal | |
Antibody Type | Primary | |
Host | Rabbit | |
Isotype | IgG | |
Western Blot | Yes | |
ImmunoChemistry | Yes | |
Antigen Synonyms | melatonin receptor MEL1B, melatonin receptor 1B, FGQTL2, MEL-1B-R, MT2, Mel 1b melatonin receptor, Mel1b receptor, melatonin receptor type 1B | melatonin receptor MEL1B, Mel 1b melatonin receptor, Mel1b receptor, melatonin receptor type 1B, melatonin receptor 1B, MT2, FGQTL2, MEL-1B-R |
Gene ID | 4544 | |
Concentration | 1 mg/ml | |
Immunogen | A synthetic peptide made to an internal region of the human Melatonin Receptor 1B protein (within residues 200-300) [Swiss-Prot P49286]. | |
Purification | Immunogen affinity purified | |
Storage Buffer | PBS and 30% Glycerol | |
Storage Temperature | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |