Anti-MTNR1B Rabbit Polyclonal Antibody

NBP1-00922
102155-050EA 466.15 USD
102155-050
Anti-MTNR1B Rabbit Polyclonal Antibody
Rabbit Polyclonal Melatonin Receptor 1B Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MTNR1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR