Anti-OPN4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-74460
102116-386EA 635.51 USD
102116-386
Anti-OPN4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Melanopsin Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen. Tested Reactivity: Mouse, Rat.

Type: Primary
Antigen: OPN4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR