Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit Polyclonal anti-Melanoma antigen family C2 Antibody, Novus Biologicals (NBP1-68961)

Supplier: Novus Biologicals
NBP1-68961
Rabbit Polyclonal Melanoma antigen family C2 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: Melanoma antigen family C2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-68961 102198-246 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol Melanoma Antigen Family C2
Antigen Name Melanoma antigen family C2
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms MAGE-C2 antigen, HCA587MAGE-C2, CT10MAGE-E1 antigen, Cancer/testis antigen 10, melanoma antigen, Hepatocellular carcinoma-associated antigen 587, family E, melanoma antigen family C, MAGEE1MGC13377, 1, 2, hepatocellular cancer antigen 587, cancer/testis specific, melanoma-associated antigen C2
Gene ID 51438
Immunogen Synthetic peptides corresponding to MAGEC2 (melanoma antigen family C, 2) The peptide sequence was selected from the N terminal of MAGEC2. Peptide sequence SCCSSFSWSSFSEESSSQKGEDTGTCQGLPDSESSFTYTLDEKVAELVEF.
Purification Immunogen affinity purified
Storage Buffer PBS and 2% Sucrose
Storage Temperature Store at -20C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty