Anti-MAP3K2 Rabbit Polyclonal Antibody

NBP1-21404
102160-304EA 536.92 USD
102160-304
Anti-MAP3K2 Rabbit Polyclonal Antibody
Rabbit Polyclonal MEKK2 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: MAP3K2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR