Anti-MAP2K4 Rabbit Polyclonal Antibody

NB100-91957
102119-622EA 466.15 USD
102119-622
Anti-MAP2K4 Rabbit Polyclonal Antibody
Rabbit Polyclonal MEK4 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR