Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MEK1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
Rabbit Polyclonal MEK1 [p Thr386] Antibody. Tested Applications: Western Blot. Tested Reactivity: Rat.

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Host: Rabbit
Reactivity: Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NB500-144 102131-844 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen Symbol MEK1
Antigen Name Mitogen-activated Protein Kinase Kinase 1
Reactivity Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms dual specificity mitogen-activated protein kinase kinase 1, PRKMK1EC, kinase 1 (MAP kinase kinase 1), MKK1MEK 1, MAPKK1, MAP kinase kinase 1, ERK activator kinase 1, mitogen-activated protein kinase kinase 1, MAPK/ERK kinase 1, protein kinase, MEK1MAPKK 1, mitogen-activated
Gene ID 5604
Modification p Thr386
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr386 conjugated to KLH
Purification Immunogen affinity purified
Storage Buffer 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol
Storage Temperature Store at -20C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty