Anti-MEK1 Rabbit Polyclonal Antibody

NBP2-12966
102252-712EA 466.15 USD
102252-712
Anti-MEK1 Rabbit Polyclonal Antibody
Rabbit Polyclonal MEK1 [p Ser298] Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR