Anti-MEF2D Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80567
102216-770EA 635.51 USD
102216-770
Anti-MEF2D Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MEF2D Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MEF2D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR