Anti-MED24 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-74599
102116-538EA 635.51 USD
102116-538
Anti-MED24 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MED24 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MED24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR