Anti-MED19 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79597
102214-842EA 564.74 USD
102214-842
Anti-MED19 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MED19 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Rat.

Type: Primary
Antigen: MED19
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR