Anti-MED10 Rabbit Polyclonal Antibody

NBP1-79402
102214-452EA 466.15 USD
102214-452
Anti-MED10 Rabbit Polyclonal Antibody
Rabbit Polyclonal MED10 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MED10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR