Anti-MDM2 Rabbit Polyclonal Antibody

41420002
102103-918EA 423.08 USD
102103-918
Anti-MDM2 Rabbit Polyclonal Antibody
Rabbit Polyclonal MDM2 Antibody. Tested Applications: ELISA. Tested Reactivity: Human.

Type: Primary
Antigen: MDM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR