Anti-MDFIC Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91377
102217-292EA 564.74 USD
102217-292
Anti-MDFIC Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MDFIC Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Rabbit.

Type: Primary
Antigen: MDFIC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR