Anti-MCSF1R Rabbit Polyclonal Antibody (DyLight® 650)

NBP1-19380C
102158-750EA 423.08 USD
102158-750
Anti-MCSF1R Rabbit Polyclonal Antibody (DyLight® 650)
Rabbit Polyclonal MCSF Receptor Antibody [DyLight 650]. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MCSF1R
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR