Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL2/MCP1 Rabbit Polyclonal Antibody (DyLight 650) [clone: ]

Supplier: Novus Biologicals
NBP1-07034C
The CCL2 / MCP1 Antibody [DyLight 650] from Novus Biologicals is a rabbit polyclonal antibody to CCL2 / MCP1. This antibody reacts with human, rat. The CCL2 / MCP1 Antibody [DyLight 650] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: CCL2/MCP1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-07034C 102157-596 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol CCL2/MCP1
Antigen Name Monocyte Chemoattractant Protein 1
Reactivity Rat, Human
Conjugation DyLight® 650
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms MGC9434, SCYA2HSMCR30, C-C motif chemokine 2, chemokine (C-C motif) ligand 2, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, small inducible cytokine subfamily A (Cys-Cys), small inducible cytokine A2 (monocyte chemotactic protein 1, GDCF-2, homologous tomouse Sig-je), MCP1 Monocyte chemotactic protein 1, member 2, MCP-1 Small-inducible cytokine A2, SMC-CF
Gene ID 6347
Immunogen Synthetic peptide made to an internal portion of rat MCP1 (within residues 30-80). [Swiss-Prot# P14844]
Purification Immunogen affinity purified
Storage Buffer 50mM Sodium Borate
Storage Temperature Store at 4C in the dark.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty