Anti-MCM8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-58129
102190-948EA 564.74 USD
102190-948
Anti-MCM8 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MCM8 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml

Type: Primary
Antigen: MCM8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR