Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCM3AP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-71883
Rabbit Polyclonal MCM3AP Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: MCM3AP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-71883 102201-640 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol MCM3AP
Antigen Name Minichromosome Maintenance Complex Component 3 Associated Protein
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
Antigen Synonyms minichromosome maintenance deficient (S. cerevisiae) 3-associated protein, MCM3 minichromosome maintenance deficient 3 (S. cerevisiae) associated protein, human mRNA for MCM3 import factor10Protein GANP, MCM3 minichromosome maintenance deficient 3 associated protein, 80 kDa MCM3-associated protein, Map80, minichromosome maintenance complex component 3 associated protein, FLJ44336, KIAA0572MAP80germinal center-associated nuclear protein, minichromosome maintenance deficient 3-associated protein, MCM3 import protein, GANPFLJ45306
Gene ID 8888
Concentration 1 mg/ml
Immunogen The immunogen for this product maps to a region between residue 1930 and 1980 of human Germinal center-Associated Nuclear Protein using the numbering given in entry NP_003897.2 (GeneID 8888).
Purification Immunogen affinity purified
Storage Buffer Tris-Citrate/Phosphate (pH 7.0 - 8.0)
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty