Anti-MCM2 Rabbit Polyclonal Antibody

NBP1-36085
102168-138EA 678.46 USD
102168-138
Anti-MCM2 Rabbit Polyclonal Antibody
Rabbit Polyclonal MCM2 Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: MCM2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR