Anti-MCM10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-82395
102216-850EA 564.74 USD
102216-850
Anti-MCM10 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MCM10 Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MCM10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR