Anti-MCHR2 Rabbit Polyclonal Antibody

NBP1-00916
102155-044EA 466.15 USD
102155-044
Anti-MCHR2 Rabbit Polyclonal Antibody
Rabbit Polyclonal MCHR2 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human.

Type: Primary
Antigen: MCHR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR