Anti-MCG10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76729
102209-140EA 564.74 USD
102209-140
Anti-MCG10 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MCG10 Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Type: Primary
Antigen: MCG10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR