Rabbit Polyclonal anti-MC5 Receptor Antibody, Novus Biologicals (NBP1-00915)

NBP1-00915
102155-042EA 466.15 USD
102155-042
Rabbit Polyclonal anti-MC5 Receptor Antibody, Novus Biologicals (NBP1-00915)
Rabbit Polyclonal MC5 Receptor Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MC5 Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR