Anti-MBD3 Rabbit Polyclonal Antibody

NBP1-40346
102169-190EA 536.92 USD
102169-190
Anti-MBD3 Rabbit Polyclonal Antibody
Rabbit Polyclonal MBD3 Antibody. Tested Applications: Immunoprecipitation, Western Blot (-). Tested Reactivity: Human.

Type: Primary
Antigen: MBD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR