Anti-MBD2 Rabbit Polyclonal Antibody

NB100-1703
102105-824EA 466.15 USD
102105-824
Anti-MBD2 Rabbit Polyclonal Antibody
Rabbit Polyclonal MBD2 Antibody. Tested Applications: Western Blot, Gel Super Shift Assays, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MBD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR