Anti-Maxi Potassium channel beta Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-33484
102167-070EA 635.51 USD
102167-070
Anti-Maxi Potassium channel beta Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Maxi Potassium channel beta Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: Maxi Potassium channel beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR