Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit Polyclonal anti-Maxi Potassium channel alpha Antibody, Novus Biologicals (NBP1-46701)

Supplier: Novus Biologicals
NBP1-46701
Rabbit Polyclonal Maxi Potassium channel alpha Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:300-1:2000

Type: Primary
Antigen: Maxi Potassium channel alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-46701 102176-466 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol Maxi Potassium channel alpha
Antigen Name Maxi Potassium channel alpha
Reactivity Mouse, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms MGC71881, k(VCA)alpha, bA205K10.1, Maxi K channel, DKFZp686K1437, Slo homolog, BKTM, stretch-activated Kca channel, BKCA alpha subunit, subfamily M, SAKCA, subfamily M subunit alpha-1, Slo1, BKCA alpha, alphamember 1, MaxiK, KCNMA, SLO-ALPHA, potassium large conductance calcium-activated channel, BK channel, calcium-activated potassium channel subunit alpha-1, hSlo, Slowpoke homolog
Gene ID 3778
Concentration LYOPH
Immunogen A synthetic peptide from 4th cytoplasmic domain of rat Potassium channel subfamily M subunit alpha 1 (KCNMA1, KCa1.1, SLO) conjugated to an immunogenic carrier protein was used as the antigen.
Purification Unpurified
Storage Buffer Whole antisera
Storage Temperature Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty