Anti-MAX binding protein Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-04052
102234-864EA 635.51 USD
102234-864
Anti-MAX binding protein Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MAX binding protein Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: MAX binding protein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR