Anti-MAVS Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-80859
102118-406EA 635.51 USD
102118-406
Anti-MAVS Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MAVS Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: MAVS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR