Anti-MAT2A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB110-94158SS NB110-94158
102126-380EA 208 USD
102126-380 102126-366
Anti-MAT2A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MAT2A Antibody. Tested Applications: Western Blot. Tested Reactivity: Human, Mouse, Rat, Bovine, Chicken, Primate, Porcine.

Type: Primary
Antigen: MAT2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR