Anti-MAST205 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-22985
102160-810EA 635.51 USD
102160-810
Anti-MAST205 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MAST205 Antibody. Tested Applications: Immunoprecipitation, Western Blot (-). Tested Reactivity: Human.

Type: Primary
Antigen: MAST205
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR