Anti-SERPINB5 Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB110-97377B
102126-476EA 614.15 USD
102126-476
Anti-SERPINB5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
The Serpin B5 / Maspin Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to Serpin B5 / Maspin. This antibody reacts with human. The Serpin B5 / Maspin Antibody [Biotin] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: SERPINB5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR