Anti-MARVELD3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59494
102192-680EA 564.74 USD
102192-680
Anti-MARVELD3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MARVELD3 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Canine.

Type: Primary
Antigen: MARVELD3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR