Anti-MARVELD1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76243
102207-258EA 564.74 USD
102207-258
Anti-MARVELD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MARVELD1 Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MARVELD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR