Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MARK2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-71889
Rabbit Polyclonal MARK2 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: MARK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μl NBP1-71889 102201-652 Each Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol MARK2
Antigen Name MAP/microtubule affinity-regulating kinase 2
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoPrecipitation Yes
Antigen Synonyms serine/threonine-protein kinase MARK2, MAP/microtubule affinity-regulating kinase 2MGC99619, PAR1 homolog, ELKL motif kinase 1, EMK-1, EC 2.7.11.1, ELKL motif kinase, Par1b, EMK1PAR-1, serine/threonine protein kinase EMK, EC 2.7.11, Ser/Thr protein kinase PAR-1B
Gene ID 2011
Concentration 1 mg/ml
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human MAP/microtubule Affinity-Regulating Kinase 2 using the numbering given in entry NP_001034558.2 (GeneID 2011).
Purification Immunogen affinity purified
Storage Buffer Tris-Citrate/Phosphate (pH 7.0 - 8.0)
Storage Temperature Store at 4C. Do not freeze.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty