Anti-MARK2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78028
102212-382EA 777.05 USD
102212-382
Anti-MARK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MARK2 [p Thr595] Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine.

Type: Primary
Antigen: MARK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR