Anti-MARCKS Rabbit Polyclonal Antibody

NBP1-00909
102155-030EA 466.15 USD
102155-030
Anti-MARCKS Rabbit Polyclonal Antibody
Rabbit Polyclonal MARCKS Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MARCKS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR