Anti-MARCH8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76959
102210-044EA 564.74 USD
102210-044
Anti-MARCH8 Rabbit Polyclonal Antibody
Antibodies
The MARCH8 Antibody from Novus Biologicals is a rabbit polyclonal antibody to 42802. This antibody reacts with human, mouse, rat. The MARCH8 Antibody has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: MARCH8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR