Anti-MAP4K6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-22988
102160-816EA 635.51 USD
102160-816
Anti-MAP4K6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MAP4K6 Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human, Mouse.

Type: Primary
Antigen: MAP4K6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR