Anti-MAP3K8 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-62178
102195-484EA 805.35 USD
102195-484
Anti-MAP3K8 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal MAP3K8 Antibody. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MAP3K8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR