Anti-MAP3K8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

NBP1-62179
102195-486EA 847.69 USD
102195-486
Anti-MAP3K8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Rabbit Polyclonal MAP3K8 Antibody [FITC]. Tested Applications: Western Blot, ELISA. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: MAP3K8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR