Anti-ELFN2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-91363
102217-264EA 564.74 USD
102217-264
Anti-ELFN2 Rabbit Polyclonal Antibody
Antibodies
The ELFN2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to ELFN2. This antibody reacts with human. The ELFN2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: ELFN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR