Anti-eIF4G1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-93317
102120-762EA 635.51 USD
102120-762
Anti-eIF4G1 Rabbit Polyclonal Antibody
Antibodies
The eIF4G1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to eIF4G1. This antibody reacts with human. The eIF4G1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: eIF4G1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR