Anti-eIF4E Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-57195
102189-290EA 564.74 USD
102189-290
Anti-eIF4E Rabbit Polyclonal Antibody
Antibodies
The eIF4E Antibody from Novus Biologicals is a rabbit polyclonal antibody to eIF4E. This antibody reacts with human, mouse. The eIF4E Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: eIF4E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR