Anti-eIF3X Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-93306
102120-744EA 635.51 USD
102120-744
Anti-eIF3X Rabbit Polyclonal Antibody
Antibodies
The eIF3X Antibody from Novus Biologicals is a rabbit polyclonal antibody to eIF3X. This antibody reacts with human, mouse. The eIF3X Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: eIF3X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR