Anti-EIF3M Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31045
102164-050EA 430.5 USD
102164-050
Anti-EIF3M Rabbit Polyclonal Antibody
Antibodies
The EIF3M Antibody from Novus Biologicals is a rabbit polyclonal antibody to EIF3M. This antibody reacts with human. The EIF3M Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: EIF3M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR