Anti-eIF3K Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-98396
102223-108EA 564.74 USD
102223-108
Anti-eIF3K Rabbit Polyclonal Antibody
Antibodies
The eIF3K Antibody from Novus Biologicals is a rabbit polyclonal antibody to eIF3K. This antibody reacts with mouse. The eIF3K Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: eIF3K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR