Anti-EIF3C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-511
102109-424EA 635.51 USD
102109-424
Anti-EIF3C Rabbit Polyclonal Antibody
Antibodies
The EIF3C Antibody from Novus Biologicals is a rabbit polyclonal antibody to EIF3C. This antibody reacts with human, mouse, rat. The EIF3C Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: EIF3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR